0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Syringe Needles
Needles, Spinocan®
Needles, Spinocan®
Needles, Spinocan®, 25, 75 mm, 0,50 mm, Orange
BRAU4505751-01
:
B. Braun
Needles, Spinocan®
BRAU4505751-01
:
B. Braun
Needles, Spinocan®
Needles, Spinocan®, 25, 75 mm, 0,50 mm, Orange
139,00 €
1 * 25 items
-
+
Colour code:
Orange
Ø:
0,50 mm
Gauge:
25
Description:
Needles, Spinocan®
Length:
75 mm
Seq:
009