Order Entry
Export
Orders LinkContactUsLinkComponent
Needles, Spinocan®

Needles, Spinocan®

Needles, Spinocan®, 22, 75 mm, 0,70 mm, Black
Catalog # BRAU4507754-13
Supplier:   B. Braun
Needles, Spinocan®
Catalog # BRAU4507754-13
Supplier:  B. Braun
Needles, Spinocan®
Needles, Spinocan®

Needles, Spinocan®, 22, 75 mm, 0,70 mm, Black

Price
139,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Specifications
  • Colour code:
    Black
  • Ø:
    0,70 mm
  • Gauge:
    22
  • Description:
    Needles, Spinocan®
  • Length:
    75 mm
  • Seq:
    007