Order Entry
Export
Orders LinkContactUsLinkComponent
 

Needles, Spinocan®

Needles, Spinocan®, 27, 88 mm, 0,40 mm, Grey
Catalog # BRAU4503902-01
Supplier:   B. Braun
Needles, Spinocan®
Catalog # BRAU4503902-01
Supplier:  B. Braun
Needles, Spinocan®
Needles, Spinocan®

Needles, Spinocan®, 27, 88 mm, 0,40 mm, Grey

Price
139,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Specifications
  • Colour code:
    Grey
  • Ø:
    0,40 mm
  • Gauge:
    27
  • Description:
    Needles, Spinocan®
  • Length:
    88 mm
  • Seq:
    012