Orders LinkContactUsLinkComponent
Various replacement parts
Stator face assembly for 0101-0921 valve, 1100, 1200, 0100-1851, AGLS
Catalog # 560-0148
Supplier:   SCIENCIX
Various replacement parts
Catalog # 560-0148
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
Stator face assembly for 0101-0921 valve, 1100, 1200, 0100-1851, AGLS
Price
282,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
  • Description:
    Stator face assembly for 0101-0921 valve
  • Seq:
    55
  • OEM manufacturer:
    AGLS
  • OEM equivalent:
    0100-1851
  • Model:
    1100, 1200