Various replacement parts
Pre-column filter insert, 2 µm, 717, 717 Plus, 2690, 2690D, 2695, 2695D, 2790, 2795, 2796, Alliance, LC Module 1, WAT005139, WATE
Catalog # 903-1328
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Pre-column filter insert, 2 µm
- Seq:43
- OEM manufacturer:WATE
- OEM equivalent:WAT005139
- Model:717, 717 Plus, 2690, 2690D, 2695, 2695D, 2790, 2795, 2796, Alliance, LC Module 1