Orders LinkContactUsLinkComponent
Various replacement parts
PEEK™ filter, 5 μm, LC-10AD, LC-10ADvp, LC-10ATvp, LC-20AD/AB, LC-20Ai, LC-20AT, LCMS-2020, LCMS-8030, LCMS-8040, LCMS-8045, LCMS-8050, 228-48607-91, SHIM
Catalog # CTSACTS-11305
Supplier:   SCIENCIX
Various replacement parts
Catalog # CTSACTS-11305
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
PEEK™ filter, 5 μm, LC-10AD, LC-10ADvp, LC-10ATvp, LC-20AD/AB, LC-20Ai, LC-20AT, LCMS-2020, LCMS-8030, LCMS-8040, LCMS-8045, LCMS-8050, 228-48607-91, SHIM
Price
26,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
  • Description:
    PEEK™ filter, 5 μm
  • Seq:
    40
  • OEM manufacturer:
    SHIM
  • OEM equivalent:
    228-48607-91
  • Model:
    LC-10AD, LC-10ADvp, LC-10ATvp, LC-20AD/AB, LC-20Ai, LC-20AT, LCMS-2020, LCMS-8030, LCMS-8040, LCMS-8045, LCMS-8050