Various replacement parts
Filter, air, chassis, ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr, 401000501, WATE
Catalog # CTSACTS-11150
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Filter, air, chassis
- Seq:23
- OEM manufacturer:WATE
- OEM equivalent:401000501
- Model:ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr