Needles replacement parts
Needle, 900 μl, 1100, 1200, G1313-87202, AGLS
Catalog # CTSACTS-11356
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Description:Needle, 900 μl
- Seq:02
- OEM manufacturer:AGLS
- OEM equivalent:G1313-87202
- Model:1100, 1200