Various replacement parts
Fingertight one piece 10 – 32 PEEK™ male nut/ferrule for 1,58 mm OD tubing, —, —, —
Catalog # CTSACTS-1818
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
- Description:Fingertight one piece 10 – 32 PEEK™ male nut/ferrule for 1,58 mm OD tubing
- Seq:26
- OEM manufacturer:—
- OEM equivalent:—
- Model:—