Orders LinkContactUsLinkComponent
Various replacement parts
Various replacement parts
Solvent inlet filter, stainless steel, 1050, 1090, 1100, 1200, 01018-60025, AGLS
Catalog # 903-1331
Supplier:   SCIENCIX
Various replacement parts
Catalog # 903-1331
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
Solvent inlet filter, stainless steel, 1050, 1090, 1100, 1200, 01018-60025, AGLS
Price
109,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
Specifications
  • Description:
    Solvent inlet filter, stainless steel
  • Seq:
    51
  • OEM manufacturer:
    AGLS
  • OEM equivalent:
    01018-60025
  • Model:
    1050, 1090, 1100, 1200