Various replacement parts
Sample loop, 10 μl, ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr, Open Architecture UPLC, 430001326, WATE
CTSACTS-A11147
: SCIENCIX
Jump to:
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
- Description:Sample loop, 10 μl
- Seq:48
- OEM manufacturer:WATE
- OEM equivalent:430001326
- Model:ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr, Open Architecture UPLC