Various replacement parts
Tubing assembly, transducer to check valve, ACQUITY H-Class QSM, ACQUITY Isocratic Solvent Mgr, ACQUITY M-Class µASM, ACQUITY M-Class µBSM, 430002357, WATE
Catalog # 560-0157
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
- Description:Tubing assembly, transducer to check valve
- Seq:68
- OEM manufacturer:WATE
- OEM equivalent:430002357
- Model:ACQUITY H-Class QSM, ACQUITY Isocratic Solvent Mgr, ACQUITY M-Class µASM, ACQUITY M-Class µBSM