Various replacement parts
Tube assembly, transducer to check valve, I2V, ACQUITY I-Class BSM, ACQUITY UPLC BSM, ACQUITY UPLC I2V BSM, 430001773, WATE
Catalog # CTSACTS-A11372
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Tube assembly, transducer to check valve, I2V
- Seq:62
- OEM manufacturer:WATE
- OEM equivalent:430001773
- Model:ACQUITY I-Class BSM, ACQUITY UPLC BSM, ACQUITY UPLC I2V BSM