Needles replacement parts
Needle, stainless steel, 30 μl, ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr, 205000362, WATE
CTSACTS-A11381
: SCIENCIX
Jump to:
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Description:Needle, stainless steel, 30 μl
- Seq:12
- OEM manufacturer:WATE
- OEM equivalent:205000362
- Model:ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr