Orders LinkContactUsLinkComponent
Various replacement parts
Solvent resevoir filter, 10 µm, 600, 610, 510, 515, 1515, 1525, 1525EF, 1525 Micro, 2535, 2545, 2555, 2690, 2690D, 2695, 2695D, 2700, Acquity, Acquity UPLC BSM, Alliance, LC Module 1, WAT025531, WATE
Catalog # 903-1330
Supplier:   SCIENCIX
Various replacement parts
Catalog # 903-1330
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
Solvent resevoir filter, 10 µm, 600, 610, 510, 515, 1515, 1525, 1525EF, 1525 Micro, 2535, 2545, 2555, 2690, 2690D, 2695, 2695D, 2700, Acquity, Acquity UPLC BSM, Alliance, LC Module 1, WAT025531, WATE
Price
55,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
  • Description:
    Solvent resevoir filter, 10 µm
  • Seq:
    52
  • OEM manufacturer:
    WATE
  • OEM equivalent:
    WAT025531
  • Model:
    600, 610, 510, 515, 1515, 1525, 1525EF, 1525 Micro, 2535, 2545, 2555, 2690, 2690D, 2695, 2695D, 2700, Acquity, Acquity UPLC BSM, Alliance, LC Module 1