Various replacement parts
Compression screw, stainless steel, 710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, WAT025313, WATE
Catalog # 560-0121
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Compression screw, stainless steel
- OEM manufacturer:WATE
- OEM equivalent:WAT025313
- Model:710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795