Order Entry
Export
Orders LinkContactUsLinkComponent
n-Heptane 95%

n-Heptane 95%

Glass bottle, 1.000 mL
Catalog # H33987.AU
Flammable
Health
Irritant
Environment
n-Heptane 95%
Catalog # H33987.AU
CAS Number:  142-82-5
n-Heptane 95%
n-Heptane 95%

Glass bottle, 1.000 mL

CAS Number:  142-82-5
Price
43,10 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

Purity: 95% CAS No.: 142-82-5 Molecular Formula: C7H16 Formula Weight: 100.21 Flash Point: -4°(25°F) Refractive Index: 1.3877 Odour: Petroleum-like Melting Point: -91° Boiling Point: 97-99° Beilstein Reference: 1730763 Merck Reference: 14,4659 UN No.: 1206 MDL No.: MFCD00009544

Synonyms:  high purity solvents
  • CAS Number:142-82-5
  • Formula:H₃C(CH₂)₅CH₃
  • MDL Number:MFCD00009544
  • Molecular Weight:100.20 g/mol
  • Merck Index:14,4659
  • Boiling Pt:97…99 °C
  • Melting Pt:–91 °C
  • Density:0.684
  • Flash Pt:–4 °C (25 °F)
  • Storage Temperature:Ambient
  • EINECS:205-563-8
  • UN:1206
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1.000 mL

Specifications