Fernbach culture flasks
2800 ml, with Delong neck
Catalog # 391-0533
Supplier: SciLabware
Jump to:
Product Details & Documents
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
- Exhibit 3 baffle indents to provide optimal and consistent agitation
- Large surface area to volume ratio
- Available in two versions, with Delong neck or screw cap
Specifications
- Type:with Delong neck
- Capacity:2800 ml
- Seq:01