Hematocrit tubes, PLASTICRIT™
Hematocrit Tubes, PLASTICRIT™ Heparin coated, 75 mm
Catalog # DRUM8-000-7520-H/5
Supplier: DRUMMOND
Jump to:
Product Details & Documents
These plastic hematocrit tubes provide the identical capillary action as glass hematocrit tubes. They eliminate the possibility of contamination due to breakage with no compromise in accuracy.
- Available as plain or heparinised
Complies to FDA, NIOSH, CDC, and OSHA safety recommendations.
Specifications
- Description:Hematocrit Tubes, PLASTICRIT™ Heparin coated
- Length:75 mm
- Seq:03