Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Lysine monohydrochloride

L(+)-Lysine monohydrochloride

Bottle, 500 g
Catalog # RC-067
Supplier:   G-Biosciences
L(+)-Lysine monohydrochloride
Catalog # RC-067
Supplier:  G-Biosciences
CAS Number:  657-27-2
L(+)-Lysine monohydrochloride

Bottle, 500 g

CAS Number:  657-27-2
Price
179,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
  • CAS Number:657-27-2
  • Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
  • MDL Number:MFCD00064564
  • Molecular Weight:182.65 g/mol
  • Storage Temperature:Freezer
  • EINECS:213-091-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications