Tricine buffer substance
Bottle, 500 g
Catalog # RC-104
Supplier: G-Biosciences
Jump to:
Product Details & Documents
This tricine is a buffer component used for the separation of low molecular weight peptides.
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
- CAS Number:5704-04-1
- Formula:(HOCH₂)₃CNHCH₂CO₂H
- MDL Number:MFCD00004277
- Molecular Weight:179.17 g/mol
- Merck Index:13,09724
- Melting Pt:187 °C
- Density:1.523 g/cm³ (19.85 °C)
- Storage Temperature:Ambient
- EINECS:227-193-6
Specifications
Specifications
- Pack type:Bottle
- Pk:500 g



