Order Entry
Export
Orders LinkContactUsLinkComponent
dNTP sets (dA, dC, dG, dT) 100 mM solutions

dNTP sets (dA, dC, dG, dT) 100 mM solutions

dATP, 100 mM solution, 0,25 ml, 25 mL
Catalog # 786-465
Supplier:   G-Biosciences
dNTP sets (dA, dC, dG, dT) 100 mM solutions
Catalog # 786-465
Supplier:  G-Biosciences
CAS Number:  
dNTP sets (dA, dC, dG, dT) 100 mM solutions
dNTP sets (dA, dC, dG, dT) 100 mM solutions

dATP, 100 mM solution, 0,25 ml, 25 mL

CAS Number:  
Price
67,50 €
Log in for availability
1 * 25 mL
Product Details & Documents

Set consists of 100 mM aqueous solutions of dATP, dCTP, dGTP and dTTP each supplied in a separate vial. Also available separately.

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    dATP, 100 mM solution, 0,25 ml

Specifications