Bath fluids, Haake
Sil 180, –40...+200 °C, >225 °C, 170 °C, 11 cP, Colourless, 5 L
Catalog # HAKA999-0203
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.
- Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
- Low corrosive tendency
The oils are not suitable for contact with silicone tubing.
Specifications
Specifications
- Pk:5 L
- Colour:Colourless
- Fire point:>225 °C
- Flash point:170 °C
- Temperature range:–40...+200 °C
- Type:Sil 180
- Viscosity:11 cP



