Interchangeable Blocks for PHMT Thermoshakers
Spare block for PSC32, 20×0,2 ml and 12×1,5 ml microtubes
Catalog # 444-0748
Supplier: GRANT INSTRUMENTS
Jump to:
Product Details & Documents
Specifications
- Description:Spare block for PSC32
- Seq:06
- For:20×0,2 ml and 12×1,5 ml microtubes



