Order Entry
Export
Orders LinkContactUsLinkComponent
 

Scrapers, SteriWare®

Sterile, 225 mm, 110×110 mm
  SAMP8785H-110S
Scrapers, SteriWare®
  SAMP8785H-110S
Scrapers, SteriWare®
Scrapers, SteriWare®

Sterile, 225 mm, 110×110 mm

 
371,00 €
   
1 * 100 items
 

These disposable scrapers are made from high quality HDPE. They are ideal in the manufacture of creams, ointments and pastes.

  • Moulded & packed in a cleanroom
  • Made from FDA and EU conforming HDPE
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10-6)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling

Full batch traceability

 : Individually bagged.

 
  • Wiping area app.:
    110×110 mm
  • Description:
    Sterile
  • Length:
    225 mm
  • Seq:
    04