Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Bottles
Glass Bottles
Winchester bottles
Winchester bottles
Print
Winchester bottles
1000 ml, Winchester bottle, Amber
Catalog # PONT010344
Supplier:
PONT PACKAGING
Winchester bottles
Catalog # PONT010344
Supplier:
PONT PACKAGING
Expand image
Return to Top
Winchester bottles
1000 ml, Winchester bottle, Amber
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 12 items
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Capacity:
1000 ml
Colour:
Amber
Description:
Winchester bottle
Seq:
02