Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Mortars, without Pestles

VWR®, Mortars, without Pestles

75 ml, 46 mm, 91 mm
Catalog # 410-0110
Supplier:   VWR Collection
VWR®, Mortars, without Pestles
Catalog # 410-0110
Supplier:  VWR Collection
VWR®, Mortars, without Pestles
VWR®, Mortars, without Pestles

75 ml, 46 mm, 91 mm

Price
21,40 €
Log in for availability
1 * 1 items
Product Details & Documents

Porcelain, produced according to DIN 12906.

  • With spout
  • Glazed outside, grinding surface unglazed
  • Highly resistant to acids and alkalis
Specifications
  • Ø int.:
    91 mm
  • Capacity:
    75 ml
  • Height:
    46 mm