Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Pestles

VWR®, Pestles

30 mm, 135 mm
Catalog # 410-0120
Supplier:   VWR Collection
VWR®, Pestles
Catalog # 410-0120
Supplier:  VWR Collection
VWR®, Pestles
VWR®, Pestles

30 mm, 135 mm

Price
16,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Porcelain.

  • Grinding surface unglazed
  • Glazed handle

Certifications: Produced according to DIN 12906.

Documents

Specifications
  • Head Ø:
    30 mm
  • Length:
    135 mm