Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Pestles

VWR®, Pestles

48 mm, 185 mm
Catalog # 410-0123
Supplier:   VWR Collection
VWR®, Pestles
Catalog # 410-0123
Supplier:  VWR Collection
VWR®, Pestles
VWR®, Pestles

48 mm, 185 mm

Price
36,10 €
Log in for availability
1 * 1 items
Product Details & Documents

Porcelain.

  • Grinding surface unglazed
  • Glazed handle

Certifications: Produced according to DIN 12906.

Documents

Specifications
  • Head Ø:
    48 mm
  • Length:
    185 mm