Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Blue, 36 (6×6), 13 mm, 106×106×57 mm
  212-0382
VWR®, Test Tubes Racks
  212-0382
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Blue, 36 (6×6), 13 mm, 106×106×57 mm

 
16,60 €
   
1 * 1 items
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    13 mm
  • No. of Holes:
    36 (6×6)
  • Colour:
    Blue
  • Seq:
    22
  • W×D×H:
    106×106×57 mm