Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Test Tubes Racks

VWR®, Test Tubes Racks

Green, 9 (3×3), 30 mm, 110,6×110,6×84,2 mm
Catalog # 212-0397
Supplier:   VWR Collection
VWR®, Test Tubes Racks
Catalog # 212-0397
Supplier:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Green, 9 (3×3), 30 mm, 110,6×110,6×84,2 mm

Price
22,10 €
Log in for availability
1 * 1 items
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    30 mm
  • No. of Holes:
    9 (3×3)
  • Colour:
    Green
  • Seq:
    02
  • W×D×H:
    110,6×110,6×84,2 mm