Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

White, 20 (4×5), 20 mm, 130×105×84 mm
  212-0393
VWR®, Test Tubes Racks
  212-0393
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

White, 20 (4×5), 20 mm, 130×105×84 mm

 
28,60 €
   
1 * 1 items
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    20 mm
  • No. of Holes:
    20 (4×5)
  • Colour:
    White
  • Seq:
    10
  • W×D×H:
    130×105×84 mm