Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Orange, 40 (4×10), 20 mm, 104×253,6×86,6 mm
  212-0415
VWR®, Test Tubes Racks
  212-0415
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Orange, 40 (4×10), 20 mm, 104×253,6×86,6 mm

 
20,60 €
   
1 * 1 items
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    20 mm
  • No. of Holes:
    40 (4×10)
  • Colour:
    Orange
  • Seq:
    34
  • W×D×H:
    104×253,6×86,6 mm