Roller bottles, Nunc™ TufRol™
Roller bottles, TufRol™, smooth surface, with easy on/off non-vented screw cap, 850 cm², 2 per pack, 20 per case
Catalog # 392-0336
Supplier: Thermo Fisher Scientific
Roller bottles, Nunc™ TufRol™
Catalog # 392-0336
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, sterile.
- One-piece seamless design reduces possibility of leaking through a seam
- Shallow and deep indents allow for excellent manual and automated handling
- Proprietary manufacturing process allows for greater impact resistance
- Vertical expanded surfaces version facilitates emptying and reduces retention of product
Nunc™ TufRol™ have been developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics, where reliability and reproducibility are essential.
Caution: The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Growth area:850 cm²
- Packed:2 per pack, 20 per case
- Description:Roller bottles, TufRol™, smooth surface, with easy on/off non-vented screw cap
- Cat. No.:392-0336
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Vented caps for TufRol™ roller bottles, HDPE, sterileVented caps for TufRol™ roller bottles, HDPE, sterile392-0326Log in for availability1 * 500 items - 549,00 €