Nutrient Pad Sets, NKS e.motion
Schaufus pottinger/
m green yeast and mold , 0,45 µm, Yeast and moulds, 100
m green yeast and mold , 0,45 µm, Yeast and moulds, 100
Catalog # SART14091-50-RDN
Supplier: Sartorius
Jump to:
Product Details & Documents
Nutrient pad sets for colony counting by the membrane filter method, specially designed for the Microsart e.motion dispenser. These nutrient pad consist of dehydrated culture media inserted into pre-sterilised Petri dishes. This alternative to agar media has a long storage life of 24 months at room temperature.
- Economical, time-saving microbiological control
- High quality and reproducible results for up to 24 months
- Compliant with international standards and references
- Easy handling
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100
- Pore size:0,45 µm
- Type:Schaufus pottinger/<br />m green yeast and mold
- For:Yeast and moulds