Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TESTOMAT 2000 CHLOR CL 2250 A 1 * 500 mL
Catalog # HEYL156230
Supplier:
Heyl GmbH & Co, Gebr.
Expand image
TESTOMAT 2000 CHLOR CL 2250 A 1 * 500 mL
Catalog # HEYL156230
Supplier:
Heyl GmbH & Co, Gebr.