Pipette tips, OneTouch™
1 - 20 µl, +, 96 tips/rack, 10 racks/pack
Catalog # SORE10310
Supplier: Sorenson Bioscience
Jump to:
Product Details & Documents
These tips are manufactured with the OneTouch™ dual shot technology. They are made from two different resins: The body is made of rigid PP while the top is made of soft TPE material.
- Rigid PP body for glass-like clarity; soft TPE top for easy sealing and ejection
- Tips reduce the necessary sealing and reciprocal ejection force felt with traditional single resin PP tips
- Soft seal offers a more universal fit: Suitable for use with most pipette brands
- Ideal for single and multi-channel pipettors
- Certified RNase/DNase-free, PCR-inhibitor-free and pyrogen-free
- Top collar is colour-coded by volume
Specifications
- Packed:96 tips/rack, 10 racks/pack
- Sterile:+
- Volume:1 - 20 µl
- Seq:02