Petri dishes
Without vents, 60 mm, 8 mm, Aseptic, 50×10/bag
: Anicrin
PS. Suitable for all microbiology applications.
- Ø:60 mm
- Packed:50×10/bag
- Sterility:Aseptic
- Version:Without vents
- Height:8 mm
- Seq:01
PS. Suitable for all microbiology applications.