Order Entry
Export
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Total Iron

QUANTOFIX® Test Strips, Total Iron

QUANTOFIX Total iron 100, 0 • 2 • 5 • 10 • 25 • 50 • 100 - mg/L - Fe²⁺ ³⁺, Colour reaction with a triazine derivate, 43×123×43 mm (package), 100 test(s), 100
Catalog # MANA91344
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Total Iron
Catalog # MANA91344
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Total Iron
QUANTOFIX® Test Strips, Total Iron

QUANTOFIX Total iron 100, 0 • 2 • 5 • 10 • 25 • 50 • 100 - mg/L - Fe²⁺ ³⁺, Colour reaction with a triazine derivate, 43×123×43 mm (package), 100 test(s), 100

CAS Number:  
Price
29,50 €
Log in for availability
1 * 100 items
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips allow the quick and easy determination of total iron in solutions. The easy dip-and-read-procedure provides reliable results without the need for training or experience. Within one minute one gets reliable results.

Iron is frequently used for water pipes and vessels in industrial applications. Therefore, dissolved iron in process water is an important indicator for the level of corrosion. In drinking water, dissolved iron is not desirable as it leads to a brown color and a foul taste. These test strips are the ideal tool to test iron concentrations in water quickly and easily down to low concentrations.

Specifications

Specifications

  • Chemical method:
    Colour reaction with a triazine derivate
  • Pk:
    100
  • Description:
    QUANTOFIX Total iron 100
  • Dimensions:
    43×123×43 mm (package)
  • Tests per kit:
    100 test(s)
  • Range:
    0 • 2 • 5 • 10 • 25 • 50 • 100 - mg/L - Fe²⁺ ³⁺
  • Cat. No.:
    MANA91344
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Iron - Triazine (white → blue-violet)
  • Weight:
    44 g

Specifications