Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichlorophenylhydrazine

2,4-Dichlorophenylhydrazine

Bottle, 1 g
Catalog # APOSOR2419-1G
Irritant
Supplier:   Apollo Scientific
2,4-Dichlorophenylhydrazine
Catalog # APOSOR2419-1G
Supplier:  Apollo Scientific
CAS Number:  13123-92-7
2,4-Dichlorophenylhydrazine

Bottle, 1 g

CAS Number:  13123-92-7
Price
138,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (2,4-Dichlorophenyl)hydrazine
  • CAS Number:13123-92-7
  • Formula:C₆H₆Cl₂N₂
  • MDL Number:MFCD00044514
  • Molecular Weight:177.031 g/mol
  • Melting Pt:91…94 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications