Orders LinkContactUsLinkComponent
Mop heads, DryMop™
Mop heads, DryMop™
Dry Mop™ frame, heavy-gauge metal with zinc-plating, 1219 mm
Catalog # 129-0606
Supplier:   MICRONOVA
Mop heads, DryMop™
Catalog # 129-0606
Supplier:  MICRONOVA
Mop heads, DryMop™
Mop heads, DryMop™
Dry Mop™ frame, heavy-gauge metal with zinc-plating, 1219 mm
Price
46,50 €
Log in for availability
1 * 1 items
Product Details & Documents
For dusting and dry mopping large surface areas.
  • Low profile design
  • Polyester pad head limits particle generation
  • Swivel head for easy maneuvering
  • Launderable/Autoclavable covers
Specifications
  • Description:
    Dry Mop™ frame, heavy-gauge metal with zinc-plating
  • Length:
    1219 mm
  • Seq:
    03