Flathead mop covers and hardware, Slimline™
NovaPoly™ mop cover, irradiated, width 230 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
Catalog # 129-0630
Supplier: MICRONOVA
Product Details & Documents
A series of elasticised slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitising all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
- Covers are easy to change and fit securely over mop frame
- Fully encased Urethane Foam layer for ample disinfectant dispersal
- Single-ply covers available without foam interior
- Fits on QPSL plastic frame or QDSL stainless steel frame
- Suitable for use with strong disinfectants and repeated autoclaving
The Micronova Slimline™ Flat Head Mop Covers fit easily and securely onto the 360° swivel SlimLine™ plastic or stainless steel mop adapters, which can be locked at 180° for more controlled cleaning.
: All components must be ordered separately.
Specifications
- Description:NovaPoly™ mop cover, irradiated, width 230 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
- Seq:04
- Cat. No.:129-0630
- Environmentally Preferable:Waste Reducing
Complementary Products
Product Information
- Catalog #DescriptionLengthMaterialAvailabilityPrice
- 129-0551Stainless steel quick disconnect extendable handle965 - 1829 mmStainless steel 304Log in for availability1 * 1 items - 358,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)129-0537Log in for availability1 * 1 items - 433,00 €
Product Information
- Catalog #L×WAvailabilityPrice
- SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handlesSlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handlesMICRQPSL-099×4" (22.9×10 cm) mmLog in for availability1 * 1 items - 151,00 €
Product Information
- Catalog #AvailabilityPrice
- MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesMicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles129-0170Log in for availability1 * 1 items - 157,00 €
Product Information
- Catalog #L×WAvailabilityPrice
- SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handlesSlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handles129-097614×4" (35.5×10 cm) mmLog in for availability1 * 1 items - 164,00 €
Product Information
- Catalog #L×WAvailabilityPrice
- SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handlesSlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handlesMICRQPSL-1818×4" (45.7×10 cm) mmLog in for availability1 * 1 items - 180,00 €
Product Information
- Catalog #L×WAvailabilityPrice
- SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handlesSlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles129-053818×4" (45.7×10 cm) mmLog in for availability1 * 1 items - 450,00 €
Product Information
- Catalog #L×WAvailabilityPrice
- SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handlesSlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles129-05369×4" (22.9×10 cm) mmLog in for availability1 * 1 items - 379,00 €
Product Information
- Catalog #L×WAvailabilityPrice
- SlimLine™ plastic mop adapters, polypropylene and polyketone material, use with 16" flat head mopsSlimLine™ plastic mop adapters, polypropylene and polyketone material, use with 16" flat head mopsMICRQPSL-16410×100 mmLog in for availability1 * 1 items - 172,00 €