Cryoboxes, Cardboard
9×9, 13 mm
Catalog # HERA820081
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Cryo resistant cardboard.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
Documents
HERA820002_TDS_EN
Datasheet
Specifications
- Compartments:9×9
- For tube Ø:13 mm