Order Entry
Export
Orders LinkContactUsLinkComponent
3-Hydroxypropionic acid 30% in aqueous solution

3-Hydroxypropionic acid 30% in aqueous solution

Bottle, 25 g
Catalog # APOSOR59874-25G
Corrosive
Irritant
Supplier:   Apollo Scientific
3-Hydroxypropionic acid 30% in aqueous solution
Catalog # APOSOR59874-25G
Supplier:  Apollo Scientific
CAS Number:  503-66-2
3-Hydroxypropionic acid 30% in aqueous solution

Bottle, 25 g

CAS Number:  503-66-2
Price
319,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Hydroxypropionic Acid, 3-Hydroxypropionic Acid (contains varying amounts of 3,3-Oxydipropionic Acid), Hydracrylic Acid
  • CAS Number:503-66-2
  • Formula:C₃H₆O₃
  • MDL Number:MFCD00058998
  • Molecular Weight:90.08 g/mol
  • Density:1.08 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
  • EINECS:207-974-8
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications