Orders LinkContactUsLinkComponent
Polishing cloths, KIMTECH PREP*
White, 600×390 mm, In bucket, 1 roll of 300 sheets
Catalog # 115-1537
Supplier:   KIMBERLY CLARK
Polishing cloths, KIMTECH PREP*
Catalog # 115-1537
Supplier:  KIMBERLY CLARK
Polishing cloths, KIMTECH PREP*
Polishing cloths, KIMTECH PREP*
White, 600×390 mm, In bucket, 1 roll of 300 sheets
Price
579,00 €
Log in for availability
1 * 1 Roll
Product Details & Documents
The KIMTECH PREP* polishing wipes are ideal spunlace products for dry polishing glass and other materials. They are also suitable for pre- and post-paint applications.
  • Large centrefeed rolls in dispensing bucket
  • Soft and bulky - no scratching
Specifications
  • Packed:
    In bucket, 1 roll of 300 sheets
  • Colour:
    White
  • L×W:
    600×390 mm