Accessories for Evolution™ UV/VIS Spectrophotometers
Smart thermostatted 7-cell changer, 15×9×10, 16 lbs., 0...95 °C temp. range, —, Accessory for evolution spectrometers
Catalog # SPET840-219000
Supplier: Thermo Fisher Scientific
Accessories for Evolution™ UV/VIS Spectrophotometers
Catalog # SPET840-219000
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
- Model:—
- Description:Smart thermostatted 7-cell changer, 15×9×10, 16 lbs., 0...95 °C temp. range
- Seq:9
- For:Accessory for evolution spectrometers