VWR®, Yeast extract
Yeast extract, 5 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
- High purity extract
- Used for supplementing media
Documents
Specifications
Specifications
- Pk:5 kg
- Description:Yeast extract