Order Entry
Export
Orders LinkContactUsLinkComponent
Acetone ≥99.5% ACS

Acetone ≥99.5% ACS

Glass bottle, 4 L
Catalog # E646-4L
Flammable
Irritant
Supplier:   VWR Chemicals
Acetone ≥99.5% ACS
Catalog # E646-4L
Supplier:  VWR Chemicals
CAS Number:  67-64-1
Acetone ≥99.5% ACS
Acetone ≥99.5% ACS

Glass bottle, 4 L

CAS Number:  67-64-1
Price
315,00 €
Log in for availability
1 * 4 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specification Test Results

  • Abs.@330 nm:
    <1
  • Abs.@340 nm:
    ≤0.1
  • Abs.@350 nm:
    ≤0.02
  • Abs.@400 nm:
    ≤0.01
  • Aldehyde:
    ≤0.002%
  • Color (APHA):
    ≤10
  • Isopropyl Alcohol:
    ≤0.05%
  • Methanol:
    ≤0.05%
  • Purity:
    ≥99.5%
  • Residue After Evaporation:
    ≤0.001%
  • Solubility (50%, CO₂ free Water):
    Pass
  • Substances Reducing Permanganate Test:
    Pass
  • Titrable Acid:
    ≤0.0003 meq/g
  • Titrable Base:
    ≤0.0006 meq/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L

Specifications