Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Tris-borate-EDTA (TBE) buffer

VWR®, Tris-borate-EDTA (TBE) buffer

TBE buffer, 10X liquid concentrate, Plastic bottle, 500 mL
Catalog # 0658-500ML
VWR®, Tris-borate-EDTA (TBE) buffer
Catalog # 0658-500ML
Supplier:  
CAS Number:  
VWR®, Tris-borate-EDTA (TBE) buffer
VWR®, Tris-borate-EDTA (TBE) buffer

TBE buffer, 10X liquid concentrate, Plastic bottle, 500 mL

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
TBE buffer (10X) Molecular biology grade 1 * 1 L Catalog #A3945.1000

Product A3945.1000 is a replacement for 0658-500ML but varies by Brand, Packaging or Quantity.

TBE 10X LIQUID CONCENTRATE 1 * 500 mL Catalog #E442-500ML

Product E442-500ML is a replacement for 0658-500ML but varies by Brand, Packaging or Quantity.

TBE 10X LIQUID CONC. ULTRA PURE GRADE 1 * 1 L Catalog #0658-1L

Product 0658-1L is a replacement for 0658-500ML but varies by Brand, Packaging or Quantity.

Product Details & Documents

TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose. Buffers are available as liquid concentrate or Ready-Pack™ (a foil pouch containing sufficient material to prepare 1 litre of 10X concentrate).

  • High buffering capacity
  • High ionic strength
  • Allows extended electrophoresis runs without buffer recirculation
  • Convenient packaging options minimise weighing and mixing
  • Free from DNase, RNase and protease

Composition of (1X) solution contains 0,089 M Tris base, 0,089 M borate and 0,002 M EDTA, disodium salt, dihydrate with a final pH of 8,3.

 : *Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    TBE buffer, 10X liquid concentrate

Specifications