Order Entry
Export
Orders LinkContactUsLinkComponent
Water for HPLC

Water for HPLC

Glass bottle, 1 L
Catalog # K978-1L
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Water for HPLC
Catalog # K978-1L
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  7732-18-5
Water for HPLC
Water for HPLC

Glass bottle, 1 L

CAS Number:  7732-18-5
Price
102,00 €
Log in for availability
1 * 1 L
Product Details & Documents

High purity water.

  • Functionally tested reagents suitable for HPLC and mass spectrometry
  • Low UV absorbing and low particulate count to reduce background
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specification Test Results

  • Abs.@200nm:
    0.1
  • Abs.@210nm:
    0.1
  • Abs.@254nm:
    0.005
  • Abs.@300nm:
    0.005
  • Ammonia:
    0.3 ppm
  • Calcium Test:
    Pass
  • Carbon Dioxide Test:
    Pass
  • Chloride:
    0.5 ppm
  • DNase:
    none detected
  • Endotoxins:
    0.25 EU/mL
  • Liquid Chromatography Suitability:
    Pass
  • Oxidizable Substances:
    Pass
  • pH @25°C:
    5.0-7.0
  • Protease:
    none detected
  • RNase:
    none detected

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    K978-1L
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications